Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 32 - 33 | 3719 | 150 | |
| GenBank mRNA | AF172276.2 | - | 32 - 33 | 3798 | 150 | |
| AK004700.1 | - | 21 - 22 | 2515 | 150 | ||
| AK030109.1 | - | 28 - 29 | 3260 | 150 | ||
| AK149434.1 | - | 32 - 33 | 3681 | 150 | ||
| AY347570.1 | - | 32 - 33 | 3599 | 150 | ||
| BC049853.1 | - | 32 - 33 | 3671 | 150 | ||
| BC138876.1 | - | 32 - 33 | 3698 | 8861898 | 150 | |
More Information
Set Membership: |
|

Aox3
Genomic Map