Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 7 - 8 | 714 | 99 | |
| GenBank mRNA | AK030109.1 | - | 3 - 4 | 253 | 100 | |
| AK149434.1 | - | 7 - 8 | 675 | 99 | ||
| BB616838.1 | - | 3 - 4 | 253 | 100 | ||
| BC024046.1 | - | 7 - 8 | 664 | 99 | ||
| BC049853.1 | - | 7 - 8 | 666 | 99 | ||
| BC138876.1 | - | 7 - 8 | 693 | 8861898 | 99 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox3
Genomic Map