Assay Details
| Gene Symbol: Aox3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023617.2 | NP_076106.2 | 8 - 9 | 795 | 89 | |
| GenBank mRNA | AK030109.1 | - | 4 - 5 | 334 | 89 | |
| AK149434.1 | - | 8 - 9 | 756 | 89 | ||
| BB616838.1 | - | 4 - 5 | 334 | 89 | ||
| BC049853.1 | - | 8 - 9 | 747 | 89 | ||
| BC138876.1 | - | 8 - 9 | 774 | 8861898 | 89 | |
More Information
Updated Mapping Information
Set Membership: |
|

Aox3
Genomic Map