Assay Details
| Gene Symbol: Mob4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025283.3 | NP_079559.2 | 5 - 6 | 411 | 70 | |
| GenBank mRNA | AK020446.1 | - | 5 - 6 | 367 | 70 | |
| AK147857.1 | - | 5 - 6 | 370 | 70 | ||
| AK162327.1 | - | 5 - 6 | 398 | 70 | ||
| BC028500.1 | - | 4 - 5 | 321 | 70 | ||
| BC037499.1 | - | 5 - 6 | 364 | 1512578 | 70 | |
| BC052475.1 | - | 5 - 6 | 356 | 70 | ||
| BC058168.1 | - | 5 - 6 | 364 | 4980300 | 70 | |
More Information
Updated Mapping Information
Set Membership: |
|

Mob4
Genomic Map