Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600025942.mm.1 |
TC0600003307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025942.mm.1 |
NM_001048054.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025942.mm.1 |
NM_130447.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025943.mm.1 |
TC0600003307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025943.mm.1 |
NM_001048054.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025943.mm.1 |
NM_130447.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025939.mm.1 |
TC0600003307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025939.mm.1 |
NM_001048054.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025939.mm.1 |
NM_130447.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025941.mm.1 |
TC0600003307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025941.mm.1 |
NM_001048054.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025941.mm.1 |
NM_130447.3 |
Refseq |