Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0600025936.mm.1 |
TC0600003307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025936.mm.1 |
NM_001048054.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025936.mm.1 |
NM_130447.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025937.mm.1 |
TC0600003307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025937.mm.1 |
NM_001048054.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025937.mm.1 |
NM_130447.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025938.mm.1 |
TC0600003307.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0600025938.mm.1 |
NM_001048054.2 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0600025938.mm.1 |
NM_130447.3 |
Refseq |