Assay Details
Gene Symbol: Dusp16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001048054.2 | NP_001041519.1 | 4 - 5 | 1248 | 80 | |
NM_130447.3 | NP_569714.2 | 4 - 5 | 1248 | 80 | ||
GenBank mRNA | AB052157.1 | - | 4 - 5 | 967 | 80 | |
AK144800.1 | - | 4 - 5 | 1249 | 80 | ||
BC057321.1 | - | 4 - 5 | 1099 | 6830600 | 80 | |
BC059232.1 | - | 4 - 5 | 1035 | 6409944 | 80 | |
CN529289.1 | - | 3 - 4 | 334 | 80 | ||
CV557369.1 | - | 3 - 4 | 524 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
