Assay Details
| Gene Symbol: Cflar | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289704.2 | NP_001276633.1 | 7 - 8 | 952 | 64 | |
| NM_001293804.1 | NP_001280733.1 | 7 - 8 | 935 | 64 | ||
| NM_001293805.1 | NP_001280734.1 | 5 - 6 | 428 | 64 | ||
| NM_207653.5 | NP_997536.1 | 7 - 8 | 1224 | 64 | ||
| GenBank mRNA | AK152515.1 | - | 7 - 8 | 919 | 64 | |
| AK161120.1 | - | 5 - 6 | 432 | 64 | ||
| BC029223.1 | - | 7 - 8 | 1193 | 4460554 | 64 | |
| U97076.1 | - | 7 - 8 | 890 | 64 | ||
| Y14041.1 | - | 6 - 7 | 802 | 64 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cflar
Genomic Map