Assay Details
| Gene Symbol: Casp8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080126.1 | NP_001073595.1 | 3 - 4 | 632 | 80 | |
| NM_001277926.1 | NP_001264855.1 | 4 - 5 | 748 | 80 | ||
| NM_009812.2 | NP_033942.1 | 3 - 4 | 805 | 80 | ||
| GenBank mRNA | AF067834.1 | - | 3 - 4 | 685 | 80 | |
| AJ000641.1 | - | 2 - 3 | 251 | 80 | ||
| AJ007749.1 | - | 3 - 4 | 458 | 80 | ||
| AK146758.1 | - | 3 - 4 | 805 | 80 | ||
| AK153340.1 | - | 3 - 4 | 514 | 80 | ||
| AK156135.1 | - | 3 - 4 | 520 | 80 | ||
| AK168812.1 | - | 3 - 4 | 805 | 80 | ||
| BC006737.1 | - | 3 - 4 | 747 | 3964119 | 80 | |
| BC049955.1 | - | 3 - 4 | 639 | 4208824 | 80 | |
| CT010166.1 | - | 2 - 3 | 412 | 80 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Casp8
Genomic Map