Assay Details
| Gene Symbol: Mthfr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161798.1 | NP_001155270.1 | 9 - 10 | 1676 | 100 | |
| NM_010840.3 | NP_034970.2 | 9 - 10 | 1599 | 100 | ||
| NR_027809.1 | 9 - 10 | 1676 | 100 | |||
| GenBank mRNA | AK030192.1 | - | 9 - 10 | 1594 | 100 | |
| AK149795.1 | - | 9 - 10 | 1661 | 100 | ||
| AK150159.1 | - | 9 - 10 | 1677 | 100 | ||
| BC051017.1 | - | 9 - 10 | 1578 | 6308248 | 100 | |
| BC052466.1 | - | 9 - 10 | 1643 | 6834886 | 100 | |
| BC100550.1 | - | 1 - 2 | 67 | 100 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Mthfr
Genomic Map