Assay Details
| Gene Symbol: Acsl3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033606.2 | NP_001028778.2 | 8 - 9 | 1471 | 153 | |
| NM_001136222.1 | NP_001129694.1 | 7 - 8 | 1053 | 153 | ||
| NM_028817.3 | NP_083093.2 | 9 - 10 | 1611 | 153 | ||
| GenBank mRNA | AK012088.1 | - | 8 - 9 | 1414 | 153 | |
| AK160839.1 | - | 7 - 8 | 997 | 153 | ||
| AK161708.1 | - | 9 - 10 | 1612 | 153 | ||
| AK166723.1 | - | 9 - 10 | 1549 | 153 | ||
| BC031529.1 | - | 7 - 8 | 1196 | 4009004 | 153 | |
More Information
Set Membership: |
|

Acsl3
Genomic Map