Assay Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278075.1 | NP_001265004.1 | 11 - 12 | 1015 | 77 | |
| NM_011811.4 | NP_035941.2 | 11 - 12 | 1015 | 77 | ||
| NR_102757.1 | 11 - 12 | 917 | 77 | |||
| NR_102758.1 | 10 - 11 | 959 | 77 | |||
| GenBank mRNA | AF123263.1 | - | 11 - 12 | 1015 | 77 | |
| AK010271.1 | - | 11 - 12 | 978 | 77 | ||
| AK045432.1 | - | 11 - 12 | 967 | 77 | ||
| AK076607.1 | - | 11 - 12 | 977 | 77 | ||
| AK134799.1 | - | 10 - 11 | 921 | 77 | ||
| AK165645.1 | - | 11 - 12 | 882 | 77 | ||
| AK167945.1 | - | 11 - 12 | 978 | 77 | ||
| AK168923.1 | - | 11 - 12 | 959 | 77 | ||
| AK169621.1 | - | 10 - 11 | 922 | 77 | ||
| AK217307.1 | - | 4 - 5 | 213 | 77 | ||
| BC002313.1 | - | 11 - 12 | 874 | 3592805 | 77 | |
| BC016428.1 | - | 11 - 12 | 982 | 4239906 | 77 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Farsb
Genomic Map