Assay Details
| Gene Symbol: Farsb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278075.1 | NP_001265004.1 | 8 - 9 | 843 | 138 | |
| NM_011811.4 | NP_035941.2 | 8 - 9 | 843 | 138 | ||
| NR_102757.1 | 8 - 9 | 745 | 138 | |||
| NR_102758.1 | 7 - 8 | 787 | 138 | |||
| GenBank mRNA | AK010271.1 | - | 8 - 9 | 806 | 138 | |
| AK012154.1 | - | 8 - 9 | 806 | 138 | ||
| AK045432.1 | - | 8 - 9 | 795 | 138 | ||
| AK076607.1 | - | 8 - 9 | 805 | 138 | ||
| AK134799.1 | - | 7 - 8 | 749 | 138 | ||
| AK167945.1 | - | 8 - 9 | 806 | 138 | ||
| AK168923.1 | - | 8 - 9 | 787 | 138 | ||
| AK169621.1 | - | 7 - 8 | 750 | 138 | ||
| CN693684.1 | - | 5 - 6 | 531 | 138 | ||
More Information
Set Membership: |
|

Farsb
Genomic Map