Assay Details
Gene Symbol: Bola1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026975.2 | NP_081251.1 | 1 - 2 | 42 | 93 | |
GenBank mRNA | AK007718.1 | - | 1 - 2 | 43 | 93 | |
BC027558.1 | - | 1 - 2 | 38 | 1477943 | 93 | |
JX420285.1 | - | 2 - 3 | 245 | 93 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AK007718.1, BC027558.1, NM_026975.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348132 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348131 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348136 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348130 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348134 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348135 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348129 | - | - |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040357 | NM_026975.2 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2040357 | AK007718.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P172014 | - | - |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040357 | NM_026975.2 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2040357 | AK007718.1 | mouse_transcriptome |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348128 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348144 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348140 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348139 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348150 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348130 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348149 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348147 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348126 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348141 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348137 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348146 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348131 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348132 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348138 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348125 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348134 | - | - |
Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348135 | - | - |
Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P172014 | - | - |
Whole Mouse Genome G4122A | A_51_P207849 | NM_026975.2 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P207849 | AK007718.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P207849 | BC027558.1 | mouse_transcriptome |
Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P172014 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02306818 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02514547 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02306817 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02514546 | - | - |
Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02306816 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348130 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348129 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348132 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348135 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348134 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348136 | - | - |
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348065 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348066 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348051 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348138 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348126 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348062 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348131 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348140 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348134 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348059 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348144 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348137 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348149 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348064 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348150 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348128 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348146 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348139 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348141 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348125 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348130 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348142 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348132 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348055 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348052 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348068 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348147 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348053 | - | - |
Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348057 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348147 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348144 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348125 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02581071 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348137 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348132 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348128 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348139 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348126 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348134 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02321447 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P02403754 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348149 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348138 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348151 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348131 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348140 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348146 | - | - |
Agilent Six_ChIP_01_of_03_v2 | A_68_P22348150 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Mouse Gene 1.0 ST Array | 10500322 | - | - |
Affymetrix Mouse Gene 1.0 ST Array | 10500323 | - | - |
Affymetrix HT MG-430 PM Array Plate | 1428456_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1452637_PM_a_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10500321 | - | - |
Mouse Genome 430 2.0 Array | 1428456_at | - | - |
Mouse Genome 430 2.0 Array | 1452637_PM_a_at | - | - |
Mouse Genome 430 2.0 Array | 1452637_a_at | - | - |
Mouse Genome 430 2.0 Array | 1428456_PM_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69168_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 69168_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR0300019501.mm.1 | NM_026975.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300019501.mm.1 | NM_026975.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300019501.mm.1 | TC0300002497.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300019502.mm.1 | NM_026975.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300019502.mm.1 | NM_026975.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300019502.mm.1 | TC0300002497.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300019499.mm.1 | NM_026975.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300019499.mm.1 | NM_026975.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300019499.mm.1 | TC0300002497.mm.1 | MTA TC |
Affymetrix Mouse Clariom S Array | PSR0300019500.mm.1 | NM_026975.2.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300019500.mm.1 | NM_026975.2 | Refseq |
Affymetrix Mouse Clariom S Array | PSR0300019500.mm.1 | TC0300002497.mm.1 | MTA TC |
Mouse Expression Set 430 | 1452637_a_at | - | - |
Mouse Expression Set 430 | 1452637_PM_a_at | - | - |
Mouse Expression Set 430 | 1428456_at | - | - |
Mouse Expression Set 430 | 1428456_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1452637_a_at | - | - |
Mouse Genome 430A 2.0 Array | 1428456_PM_at | - | - |
Mouse Genome 430A 2.0 Array | 1428456_at | - | - |
Mouse Genome 430A 2.0 Array | 1452637_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452637_PM_a_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428456_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1428456_PM_at | - | - |
Affymetrix HT Mouse Genome 430A Array | 1452637_a_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Mouse Genome Survey Microarray | 929933 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2640140 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2640140 | - | - |
