Assay Details
| Gene Symbol: Ctif | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_201354.2 | NP_958742.2 | 4 - 5 | 658 | 82 | |
| GenBank mRNA | BC058104.1 | - | 4 - 5 | 516 | 6833739 | 82 |
More Information
Updated Mapping Information
Set Membership: |
|

Ctif
Genomic Map