Assay Details
| Gene Symbol: Ciart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033302.2 | NP_001028474.1 | 4 - 5 | 640 | 72 | |
| GenBank mRNA | AK156084.1 | - | 3 - 4 | 1182 | 72 | |
| AK164001.1 | - | 4 - 5 | 640 | 72 | ||
| BC132471.1 | - | 3 - 4 | 536 | 40130748 | 72 | |
| BC138510.1 | - | 3 - 4 | 536 | 8861530 | 72 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

Ciart
Genomic Map