Assay Details
| Gene Symbol: Ciart | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033302.2 | NP_001028474.1 | 5 - 6 | 749 | 75 | |
| GenBank mRNA | AK156084.1 | - | 4 - 5 | 1291 | 75 | |
| AK164001.1 | - | 5 - 6 | 749 | 75 | ||
| BC132471.1 | - | 4 - 5 | 645 | 40130748 | 75 | |
| BC138510.1 | - | 4 - 5 | 645 | 8861530 | 75 | |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346164 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346160 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346162 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346158 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346155 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346154 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346161 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346163 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63343 | AK164001.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63343 | AK156084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63343 | NM_001033302.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63343 | AK164001.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63343 | AK156084.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P63343 | NM_001033302.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 283977 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 335204 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63343 | AK164001.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63343 | AK156084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63343 | NM_001033302.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63343 | AK164001.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63343 | AK156084.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P63343 | NM_001033302.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346156 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346155 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346162 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346167 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346140 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22346160 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04996240 | - | - |
| Whole Mouse Genome G4122A | A_52_P63343 | AK164001.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63343 | AK156084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63343 | NM_001033302.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63343 | AK164001.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63343 | AK156084.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P63343 | NM_001033302.2 | mouse_transcriptome |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346164 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346155 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346158 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346162 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346161 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346156 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346154 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346163 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22346160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346090 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346162 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346137 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346107 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346140 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346104 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346174 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346155 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346165 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346215 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346211 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22346158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346175 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346171 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02667762 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741237 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346134 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346142 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346153 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02667763 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02851040 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346137 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346146 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346138 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346160 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346169 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741242 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346168 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346155 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741232 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346174 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346140 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346162 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02741235 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22346157 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10500275 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500273 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10373453 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10500274 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1435188_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10373452 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10500272 | - | - |
| Mouse Genome 430 2.0 Array | 1435188_at | - | - |
| Mouse Genome 430 2.0 Array | 1435188_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229599_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 229599_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300019392.mm.1 | NM_001033302.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019392.mm.1 | NM_001033302.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019392.mm.1 | TC0300002489.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019390.mm.1 | NM_001033302.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019390.mm.1 | NM_001033302.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019390.mm.1 | TC0300002489.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019389.mm.1 | NM_001033302.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019389.mm.1 | NM_001033302.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019389.mm.1 | TC0300002489.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300019388.mm.1 | NM_001033302.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019388.mm.1 | NM_001033302.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300019388.mm.1 | TC0300002489.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1435188_at | - | - |
| Mouse Expression Set 430 | 1435188_PM_at | - | - |
| Murine Genome U74v2 Set | 138568_PM_at | - | - |
| Murine Genome U74v2 Set | 138568_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435188_at | - | - |
| Affymetrix HT Mouse Genome 430B Array | 1435188_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 897336 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1236079 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3029489 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_3102736 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3029489 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3102736 | - | - |

Ciart
Genomic Map