Assay Details
| Gene Symbol: Me2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145494.2 | NP_663469.1 | 2 - 3 | 211 | 66 | |
| GenBank mRNA | AK042289.1 | - | 2 - 3 | 258 | 66 | |
| AK043103.1 | - | 2 - 3 | 249 | 66 | ||
| AK050933.1 | - | 2 - 3 | 258 | 66 | ||
| AK050980.1 | - | 2 - 3 | 258 | 66 | ||
| AK077746.1 | - | 2 - 3 | 258 | 66 | ||
| AK080403.1 | - | 2 - 3 | 269 | 66 | ||
| AK156403.1 | - | 2 - 3 | 258 | 66 | ||
| AK162126.1 | - | 2 - 3 | 270 | 66 | ||
| AK171157.1 | - | 2 - 3 | 258 | 66 | ||
| BC004709.1 | - | 2 - 3 | 211 | 3499365 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

Me2
Genomic Map