Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100021369.mm.1 |
NM_007936.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021369.mm.1 |
NM_007936.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021369.mm.1 |
TC0100002618.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021368.mm.1 |
NM_007936.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021368.mm.1 |
NM_007936.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021368.mm.1 |
TC0100002618.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021370.mm.1 |
NM_007936.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021370.mm.1 |
NM_007936.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021370.mm.1 |
TC0100002618.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021367.mm.1 |
NM_007936.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021367.mm.1 |
NM_007936.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021367.mm.1 |
TC0100002618.mm.1 |
MTA TC |