Assay Details
| Gene Symbol: Plaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172695.2 | NP_766283.2 | 1 | 170 | ||
| GenBank mRNA | BC139355.1 | - | 1 | 8862377 | 170 | |
| BC139356.1 | - | 1 | 8862378 | 170 | ||
| CA490654.1 | - | 1 | 170 | |||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC139355.1,BC139356.1,NM_172695.2 transcripts.
This assay is in the 5'UTR region of the BC139355.1,BC139356.1,NM_172695.2 transcripts.
Set Membership: |
|

Plaa
Genomic Map