Assay Details
| Gene Symbol: Plaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172695.2 | NP_766283.2 | 1 - 2 | 370 | 96 | |
| GenBank mRNA | AK019070.1 | - | 1 - 2 | 375 | 96 | |
| AK075934.1 | - | 1 - 2 | 302 | 96 | ||
| AK081641.1 | - | 1 - 2 | 260 | 96 | ||
| AK162534.1 | - | 1 - 2 | 271 | 96 | ||
| AK166122.1 | - | 1 - 2 | 275 | 96 | ||
| BC139355.1 | - | 1 - 2 | 369 | 8862377 | 96 | |
| BC139356.1 | - | 1 - 2 | 369 | 8862378 | 96 | |
| BC139773.1 | - | 1 - 2 | 280 | 40111789 | 96 | |
| CA490654.1 | - | 1 - 2 | 407 | 96 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Plaa
Genomic Map