Assay Details
| Gene Symbol: Plaa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_172695.2 | NP_766283.2 | 8 - 9 | 1414 | 97 | |
| GenBank mRNA | AK019070.1 | - | 8 - 9 | 1420 | 97 | |
| AK075934.1 | - | 8 - 9 | 1346 | 97 | ||
| AK166122.1 | - | 8 - 9 | 1320 | 97 | ||
| BC139355.1 | - | 8 - 9 | 1413 | 8862377 | 97 | |
| BC139356.1 | - | 8 - 9 | 1413 | 8862378 | 97 | |
| BC139773.1 | - | 8 - 9 | 1324 | 40111789 | 97 | |
More Information
Updated Mapping Information
Set Membership: |
|

Plaa
Genomic Map