Assay Details
Gene Symbol: Mysm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177239.2 | NP_796213.2 | 17 - 18 | 2165 | 112 | |
GenBank mRNA | AK173300.1 | - | 14 - 15 | 1907 | 112 | |
BC150946.1 | - | 17 - 18 | 2161 | 9087857 | 112 | |
BC151172.1 | - | 17 - 18 | 2161 | 9088061 | 112 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
