Assay Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025728.4 | NP_080004.1 | 2 - 3 | 285 | 125 | |
| NM_029160.3 | NP_083436.2 | 11 - 12 | 1126 | 125 | ||
| GenBank mRNA | AF490391.1 | - | 11 - 12 | 1108 | 125 | |
| AK014865.1 | - | 2 - 3 | 280 | 125 | ||
| AK015884.1 | - | 11 - 12 | 1127 | 125 | ||
| AK016352.1 | - | 2 - 3 | 253 | 125 | ||
| AK016618.1 | - | 2 - 3 | 272 | 125 | ||
| BC120668.1 | - | 11 - 12 | 1125 | 40129591 | 125 | |
| BC145006.1 | - | 11 - 12 | 1125 | 9053554 | 125 | |
More Information
Updated Mapping Information
Set Membership: |
|

Spag16
Genomic Map