Assay Details
| Gene Symbol: Spag16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025728.4 | NP_080004.1 | 5 - 6 | 748 | 81 | |
| NM_029160.3 | NP_083436.2 | 14 - 15 | 1589 | 81 | ||
| GenBank mRNA | AF490391.1 | - | 14 - 15 | 1571 | 81 | |
| AK014865.1 | - | 5 - 6 | 743 | 81 | ||
| AK016618.1 | - | 5 - 6 | 735 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Spag16
Genomic Map