Assay Details
| Gene Symbol: Ercc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011729.2 | NP_035859.2 | 10 - 11 | 2634 | 78 | |
| GenBank mRNA | AK137563.1 | - | 11 - 12 | 2466 | 78 | |
| AK140825.1 | - | 10 - 11 | 2463 | 78 | ||
| AK160581.1 | - | 4 - 5 | 1508 | 78 | ||
| AK160645.1 | - | 8 - 9 | 2006 | 78 | ||
| AK164205.1 | - | 10 - 11 | 78 | |||
| BC033413.1 | - | 10 - 11 | 2354 | 78 | ||
| BC060659.1 | - | 9 - 10 | 2765 | 78 | ||
| BC092235.1 | - | 10 - 11 | 2425 | 78 | ||
| BC141070.1 | - | 10 - 11 | 2464 | 9055682 | 78 | |
| D16306.1 | - | 10 - 11 | 2329 | 78 | ||
| U40796.1 | - | 10 - 11 | 2530 | 78 | ||
More Information
Set Membership: |
|

Ercc5
Genomic Map