Assay Details
| Gene Symbol: Ercc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011729.2 | NP_035859.2 | 8 - 9 | 2267 | 65 | |
| GenBank mRNA | AK140825.1 | - | 8 - 9 | 2096 | 65 | |
| AK160581.1 | - | 2 - 3 | 1141 | 65 | ||
| AK160645.1 | - | 6 - 7 | 1639 | 65 | ||
| AK164205.1 | - | 8 - 9 | 2085 | 65 | ||
| BC033413.1 | - | 8 - 9 | 1987 | 65 | ||
| BC053034.1 | - | 8 - 9 | 2076 | 65 | ||
| BC060659.1 | - | 8 - 9 | 2066 | 65 | ||
| BC092235.1 | - | 8 - 9 | 2058 | 65 | ||
| BC141070.1 | - | 8 - 9 | 2097 | 9055682 | 65 | |
| D16306.1 | - | 8 - 9 | 1962 | 65 | ||
| U40796.1 | - | 8 - 9 | 2163 | 65 | ||
More Information
Set Membership: |
|

Ercc5
Genomic Map