Assay Details
| Gene Symbol: Map2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001039934.1 | NP_001035023.1 | 4 - 5 | 349 | 110 | |
| NM_008632.2 | NP_032658.2 | 4 - 5 | 349 | 110 | ||
| GenBank mRNA | AK046341.1 | - | 2 - 3 | 214 | 110 | |
| AK079618.1 | - | 2 - 3 | 376 | 110 | ||
| AK086484.1 | - | 2 - 3 | 213 | 110 | ||
| AK141458.1 | - | 2 - 3 | 114 | 110 | ||
| AK166380.1 | - | 4 - 5 | 365 | 110 | ||
| BC052446.1 | - | 4 - 5 | 349 | 110 | ||
| BC150945.1 | - | 4 - 5 | 348 | 9087856 | 110 | |
| BU056737.1 | - | 4 - 5 | 349 | 110 | ||
| M21041.1 | - | 2 - 3 | 115 | 110 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK141458.1, AK166380.1, BC052446.1, BC150945.1, M21041.1, NM_001039934.1, NM_008632.2 transcripts.
Set Membership: |
|

Map2
Genomic Map