Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100018523.mm.1 |
NM_028696.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018523.mm.1 |
NM_028696.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018523.mm.1 |
TC0100002305.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018524.mm.1 |
NM_028696.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018524.mm.1 |
NM_028696.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018524.mm.1 |
TC0100002305.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018519.mm.1 |
NM_028696.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018519.mm.1 |
NM_028696.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018519.mm.1 |
TC0100002305.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018522.mm.1 |
NM_028696.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018522.mm.1 |
NM_028696.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018522.mm.1 |
TC0100002305.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100018525.mm.1 |
NM_028696.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018525.mm.1 |
NM_028696.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100018525.mm.1 |
TC0100002305.mm.1 |
MTA TC |