| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0400006524.mm.1 |
NM_001122953.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006524.mm.1 |
NM_001122953.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006524.mm.1 |
NM_010905.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006524.mm.1 |
NM_010905.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006524.mm.1 |
TC0400004186.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006526.mm.1 |
NM_001122953.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006526.mm.1 |
NM_001122953.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006526.mm.1 |
NM_010905.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006526.mm.1 |
NM_010905.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006526.mm.1 |
TC0400004186.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006528.mm.1 |
NM_001122953.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006528.mm.1 |
NM_001122953.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006528.mm.1 |
NM_010905.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006528.mm.1 |
NM_010905.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006528.mm.1 |
TC0400004186.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006525.mm.1 |
NM_001122953.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006525.mm.1 |
NM_001122953.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006525.mm.1 |
NM_010905.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006525.mm.1 |
NM_010905.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006525.mm.1 |
TC0400004186.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0400006529.mm.1 |
NM_001122953.1.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006529.mm.1 |
NM_001122953.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006529.mm.1 |
NM_010905.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006529.mm.1 |
NM_010905.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0400006529.mm.1 |
TC0400004186.mm.1 |
MTA TC |