Assay Details
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122952.1 | NP_001116424.1 | 6 - 7 | 1049 | 62 | |
NM_010905.3 | NP_035035.1 | 6 - 7 | 1288 | 62 | ||
GenBank mRNA | AF326553.1 | - | 6 - 7 | 987 | 62 | |
AF326554.1 | - | 6 - 7 | 987 | 62 | ||
AK050320.1 | - | 5 - 6 | 605 | 62 | ||
AK052204.1 | - | 6 - 7 | 1110 | 62 | ||
AK054310.1 | - | 6 - 7 | 1152 | 62 | ||
AK137731.1 | - | 6 - 7 | 1151 | 62 | ||
AK139357.1 | - | 6 - 7 | 1050 | 62 | ||
AK173173.1 | - | 5 - 6 | 924 | 62 | ||
BC075702.1 | - | 6 - 7 | 1269 | 30087137 | 62 | |
D90172.1 | - | 6 - 7 | 1218 | 62 | ||
D90173.1 | - | 6 - 7 | 1126 | 62 | ||
D90174.1 | - | 6 - 7 | 1159 | 62 | ||
D90175.1 | - | 1003 | 62 | |||
U57633.1 | - | 6 - 7 | 943 | 62 | ||
Y07690.1 | - | 6 - 7 | 1019 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
