Assay Details
Gene Symbol: Nfia | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122952.1 | NP_001116424.1 | 8 - 9 | 1357 | 80 | |
NM_001122953.1 | NP_001116425.1 | 7 - 8 | 1467 | 80 | ||
NM_010905.3 | NP_035035.1 | 8 - 9 | 1596 | 80 | ||
GenBank mRNA | AF326553.1 | - | 8 - 9 | 1295 | 80 | |
AF326554.1 | - | 8 - 9 | 1295 | 80 | ||
AK052204.1 | - | 8 - 9 | 1418 | 80 | ||
AK054310.1 | - | 8 - 9 | 1460 | 80 | ||
AK137731.1 | - | 8 - 9 | 1459 | 80 | ||
AK139357.1 | - | 8 - 9 | 1358 | 80 | ||
AK173173.1 | - | 7 - 8 | 1232 | 80 | ||
BC075702.1 | - | 8 - 9 | 1577 | 30087137 | 80 | |
BC122878.1 | - | 7 - 8 | 1189 | 40107695 | 80 | |
BC122880.1 | - | 7 - 8 | 1187 | 80 | ||
D90172.1 | - | 8 - 9 | 1526 | 80 | ||
D90173.1 | - | 8 - 9 | 1434 | 80 | ||
D90174.1 | - | 8 - 9 | 1467 | 80 | ||
D90175.1 | - | 1311 | 80 | |||
U57633.1 | - | 8 - 9 | 1251 | 80 | ||
Y07690.1 | - | 8 - 9 | 1327 | 80 | ||
Y07691.1 | - | 7 - 8 | 1196 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
