Assay Details
| Gene Symbol: Grk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018869.3 | NP_061357.3 | 11 - 12 | 1895 | 61 | |
| GenBank mRNA | AF040746.1 | - | 11 - 12 | 1305 | 61 | |
| AK135076.1 | - | 11 - 12 | 1897 | 61 | ||
| AK156465.1 | - | 10 - 11 | 2270 | 61 | ||
| AK161821.1 | - | 11 - 12 | 1375 | 61 | ||
| BC019379.1 | - | 11 - 12 | 1168 | 5116510 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Grk5
Genomic Map