Assay Details
| Gene Symbol: Grk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_018869.3 | NP_061357.3 | 3 - 4 | 1094 | 70 | |
| GenBank mRNA | AF040746.1 | - | 3 - 4 | 504 | 70 | |
| AK135076.1 | - | 3 - 4 | 1096 | 70 | ||
| AK156465.1 | - | 3 - 4 | 571 | 70 | ||
| AK161821.1 | - | 3 - 4 | 574 | 70 | ||
| BC019379.1 | - | 3 - 4 | 367 | 5116510 | 70 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BI134752.1 | - | Chr. - - [-] on Build | 70 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Grk5
Genomic Map