Assay Details
| Gene Symbol: Nab1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_008667.3 | NP_032693.2 | 3 - 4 | 1208 | 119 | |
| GenBank mRNA | AB221677.1 | - | 2 - 3 | 950 | 119 | |
| BC005627.1 | - | 3 - 4 | 1208 | 3155795 | 119 | |
| BC016886.1 | - | 3 - 4 | 1131 | 4236565 | 119 | |
| U47008.1 | - | 3 - 4 | 1086 | 119 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Nab1
Genomic Map