Assay Details
| Gene Symbol: Stat1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205313.1 | NP_001192242.1 | 13 - 14 | 1428 | 120 | |
| NM_001205314.1 | NP_001192243.1 | 13 - 14 | 1464 | 120 | ||
| NM_009283.4 | NP_033309.3 | 13 - 14 | 1479 | 120 | ||
| GenBank mRNA | AK039458.1 | - | 13 - 14 | 1480 | 120 | |
| AK046517.1 | - | 13 - 14 | 1428 | 120 | ||
| AK082706.1 | - | 13 - 14 | 1461 | 120 | ||
| AK084855.1 | - | 13 - 14 | 1413 | 120 | ||
| AK157263.1 | - | 13 - 14 | 1328 | 120 | ||
| AK159887.1 | - | 13 - 14 | 1463 | 120 | ||
| BC004808.1 | - | 13 - 14 | 3587831 | 120 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Stat1
Genomic Map