Assay Details
Gene Symbol: Rnf165 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001164504.1 | NP_001157976.1 | 7 - 8 | 960 | 95 | |
GenBank mRNA | AK133955.1 | - | 4 - 5 | 347 | 95 | |
BB619563.1 | - | 4 - 5 | 347 | 95 | ||
CF745657.1 | - | 2 - 3 | 134 | 95 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
axonogenesis
motor neuron axon guidance
multicellular organism aging
protein catabolic process
positive regulation of BMP signaling pathway
forelimb morphogenesis
limb development
innervation
muscle structure development
protein binding
ubiquitin protein ligase activity
protein polyubiquitination
axonogenesis
motor neuron axon guidance
multicellular organism aging
protein catabolic process
positive regulation of BMP signaling pathway
forelimb morphogenesis
limb development
innervation
muscle structure development
protein binding
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P220262 | BC066016.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P220262 | NM_001164504.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P220262 | AK013591.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P220262 | BC066016.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P220262 | NM_001164504.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P220262 | AK013591.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117664 | AK133955.1 | mouse_transcriptome |
Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2117664 | NM_001164504.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117664 | AK133955.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2117664 | NM_001164504.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P220262 | BC066016.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P220262 | NM_001164504.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P220262 | AK013591.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P220262 | BC066016.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P220262 | NM_001164504.1 | mouse_transcriptome |
SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P220262 | AK013591.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P236629 | BC066016.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P236629 | AK036050.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_52_P236629 | NM_001164504.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P220262 | BC066016.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P220262 | NM_001164504.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P220262 | AK013591.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P220262 | BC066016.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P220262 | NM_001164504.1 | mouse_transcriptome |
Whole Mouse Genome G4122A | A_51_P220262 | AK013591.1 | mouse_transcriptome |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13818767 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P14045377 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847991 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848016 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847967 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847958 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847984 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13810314 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847969 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847951 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847966 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847946 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848005 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847968 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13818771 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847992 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848000 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13893371 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13818757 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13996404 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848001 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13893370 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13996412 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847996 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847953 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13976733 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847988 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13818769 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847972 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847970 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848019 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847965 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847975 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847964 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847976 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848002 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P13810316 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847986 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848003 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847948 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847998 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847979 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848008 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848014 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847973 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848020 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847990 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31847994 | - | - |
Agilent Six_ChIP_03_of_03_v2 | A_68_P31848011 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix HT MG-430 PM Array Plate | 1458303_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1438829_PM_at | - | - |
Affymetrix HT MG-430 PM Array Plate | 1436114_PM_at | - | - |
Affymetrix Mouse Gene 1.1 ST Array | 10459827 | - | - |
Mouse Genome 430 2.0 Array | 1438829_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1458303_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1458303_at | - | - |
Mouse Genome 430 2.0 Array | 1447404_at | - | - |
Mouse Genome 430 2.0 Array | 1447404_PM_at | - | - |
Mouse Genome 430 2.0 Array | 1438829_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225743_at | - | - |
Affymetrix GeneChip Mouse Genome 430 2.0 Array | 225743_PM_at | - | - |
Affymetrix Mouse Clariom S Array | PSR1800011694.mm.1 | NM_001164504.1.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800011694.mm.1 | NM_001164504.1 | Refseq |
Affymetrix Mouse Clariom S Array | PSR1800011694.mm.1 | TC1800001625.mm.1 | MTA TC |
Affymetrix HT Mouse Genome 430B Array | 1458303_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447404_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436114_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438829_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1438829_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1458303_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1447404_PM_at | - | - |
Affymetrix HT Mouse Genome 430B Array | 1436114_PM_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina mouse-6 v1.1 expression beadchip | ILMN_2590554 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_1259244 | - | - |
Illumina mouse-6 v1.1 expression beadchip | ILMN_2726781 | - | - |
