Assay Details
| Gene Symbol: Pstpip2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013831.4 | NP_038859.3 | 677 | 71 | ||
| GenBank mRNA | AK132418.1 | - | 659 | 71 | ||
| AK154284.1 | - | 644 | 71 | |||
| AK154724.1 | - | 7 - 8 | 683 | 71 | ||
| AK157994.1 | - | 7 - 8 | 661 | 71 | ||
| BC002123.1 | - | 658 | 3494357 | 71 | ||
| BC057654.1 | - | 619 | 4460817 | 71 | ||
| CT010308.1 | - | 7 - 8 | 509 | 71 | ||
| Y18101.1 | - | 7 - 8 | 509 | 71 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pstpip2
Genomic Map