Assay Details
| Gene Symbol: Rasef | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001017427.1 | NP_001017427.1 | 8 - 9 | 908 | 70 | |
| GenBank mRNA | AK132691.1 | - | 8 - 9 | 888 | 70 | |
| BC030882.1 | - | 4 - 5 | 305 | 70 | ||
| BC040766.1 | - | 4 - 5 | 294 | 70 | ||
| BC125539.1 | - | 7 - 8 | 722 | 40130054 | 70 | |
| BC137774.1 | - | 7 - 8 | 722 | 8860793 | 70 | |
| BC144901.1 | - | 7 - 8 | 722 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Rasef
Genomic Map