Assay Details
| Gene Symbol: Scp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011327.4 | NP_035457.1 | 2 - 3 | 231 | 79 | |
| GenBank mRNA | AK004860.2 | - | 2 - 3 | 161 | 79 | |
| BC018384.1 | - | 2 - 3 | 186 | 3710340 | 79 | |
| BC034613.1 | - | 2 - 3 | 152 | 5123611 | 79 | |
| DN176201.1 | - | 2 - 3 | 231 | 79 | ||
| M91458.1 | - | 2 - 3 | 146 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Scp2
Genomic Map