Assay Details
| Gene Symbol: Hist2h3c1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC094041.1 | - | 757 | 5099464 | 80 | |
More Information
Additional Information:
				    	 
					    	
					        	
					            	Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
					        	
					    	            
				    	
			    	
			    
			Set Membership: | 
		   
					
						
							 | 
				
Panther Classification:
MicroArray Probes:
					
						
							Agilent
					
						
							Affymetrix
					
						
							Illumina
					
					
					
						
							
								
						
					
						
							
								
						
					
						
							
								
						
					
				
			| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348404 | - | - | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094966 | AK134918.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094966 | BC139252.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094966 | NM_178215.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094966 | BC139251.1 | mouse_transcriptome | 
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2094966 | BC080809.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094966 | AK134918.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094966 | BC139252.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094966 | NM_178215.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094966 | BC139251.1 | mouse_transcriptome | 
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2094966 | BC080809.1 | mouse_transcriptome | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348417 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348416 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348414 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348410 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348411 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348408 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348406 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348404 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348407 | - | - | 
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22348413 | - | - | 
| Whole Mouse Genome G4122A | A_51_P359462 | AK134918.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P359462 | BC139252.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P359462 | NM_178215.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P359462 | BC139251.1 | mouse_transcriptome | 
| Whole Mouse Genome G4122A | A_51_P359462 | BC132490.1 | mouse_transcriptome | 
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22348404 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348455 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348322 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348381 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348419 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348416 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348457 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348453 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348414 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348384 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348439 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348433 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348451 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348417 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348377 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348438 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348373 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348458 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348328 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348386 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348333 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348437 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348274 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348410 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348272 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348353 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348431 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348325 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348440 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348355 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348362 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348413 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348420 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348408 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348406 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348321 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348375 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348275 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348380 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348378 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348330 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348371 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348351 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348450 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348435 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348323 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348359 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348332 | - | - | 
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22348368 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348420 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348411 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348438 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348430 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348412 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348407 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348431 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348416 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348437 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348423 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348404 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348406 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348439 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348435 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348424 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348417 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348429 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348410 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02759381 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348408 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348414 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02972890 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348418 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348440 | - | - | 
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22348433 | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10494403 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10494406 | - | - | 
| Affymetrix Mouse Gene 1.0 ST Array | 10494400 | - | - | 
| Affymetrix HT MG-430 PM Array Plate | 1442051_PM_at | - | - | 
| Affymetrix Mouse Gene 1.1 ST Array | 10494402 | - | - | 
| Affymetrix Mouse Gene 1.1 ST Array | 10494395 | - | - | 
| Affymetrix Mouse Gene 1.1 ST Array | 10494405 | - | - | 
| Mouse Genome 430 2.0 Array | 1442051_PM_at | - | - | 
| Mouse Genome 430 2.0 Array | 1442051_at | - | - | 
| Mouse Genome 430 2.0 Array | 1460314_PM_s_at | - | - | 
| Mouse Genome 430 2.0 Array | 1460314_s_at | - | - | 
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15077_PM_at | - | - | 
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 15077_at | - | - | 
| Affymetrix Mouse Clariom S Array | PSR0300007465.mm.1 | NM_178216.3 | Refseq | 
| Affymetrix Mouse Clariom S Array | PSR0300007465.mm.1 | TC0300003216.mm.1 | MTA TC | 
| Mouse Expression Set 430 | 1427864_at | - | - | 
| Mouse Expression Set 430 | 1453573_PM_at | - | - | 
| Mouse Expression Set 430 | 1453573_at | - | - | 
| Mouse Expression Set 430 | 1418367_PM_x_at | - | - | 
| Mouse Expression Set 430 | 1427864_PM_at | - | - | 
| Mouse Expression Set 430 | 1460314_PM_s_at | - | - | 
| Mouse Expression Set 430 | 1418366_PM_at | - | - | 
| Mouse Expression Set 430 | 1418366_at | - | - | 
| Mouse Expression Set 430 | 1460314_s_at | - | - | 
| Mouse Expression Set 430 | 1418367_x_at | - | - | 
| Mouse Expression Set 430 | 1442051_at | - | - | 
| Mouse Expression Set 430 | 1442051_PM_at | - | - | 
| Murine Genome U74v2 Set | 93022_i_at | - | - | 
| Murine Genome U74v2 Set | 93023_PM_f_at | - | - | 
| Murine Genome U74v2 Set | 93022_PM_i_at | - | - | 
| Murine Genome U74v2 Set | 93067_PM_f_at | - | - | 
| Murine Genome U74v2 Set | 96416_f_at | - | - | 
| Murine Genome U74v2 Set | 93067_f_at | - | - | 
| Murine Genome U74v2 Set | 93068_PM_r_at | - | - | 
| Murine Genome U74v2 Set | 93068_r_at | - | - | 
| Murine Genome U74v2 Set | 96416_PM_f_at | - | - | 
| Murine Genome U74v2 Set | 93023_f_at | - | - | 
| Murine Genome U74v2 Set | 94756_at | - | - | 
| Murine Genome U74v2 Set | 94756_PM_at | - | - | 
| Mouse Genome 430A 2.0 Array | 1418367_x_at | - | - | 
| Mouse Genome 430A 2.0 Array | 1427864_at | - | - | 
| Mouse Genome 430A 2.0 Array | 1453573_at | - | - | 
| Mouse Genome 430A 2.0 Array | 1460314_s_at | - | - | 
| Mouse Genome 430A 2.0 Array | 1418366_at | - | - | 
| Mouse Genome 430A 2.0 Array | 1427864_PM_at | - | - | 
| Mouse Genome 430A 2.0 Array | 1460314_PM_s_at | - | - | 
| Mouse Genome 430A 2.0 Array | 1418366_PM_at | - | - | 
| Mouse Genome 430A 2.0 Array | 1418367_PM_x_at | - | - | 
| Mouse Genome 430A 2.0 Array | 1453573_PM_at | - | - | 
| Array Name | Probe IDs | Targets | Target Type | 
|---|---|---|---|
| MouseRef-8 v2 Expression BeadChip | ILMN_3040069 | - | - | 
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3040069 | - | - | 
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_3136540 | - | - | 

Hist2h3c1
Genomic Map