Assay Details
| Gene Symbol: Gpr89 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026229.4 | NP_080505.1 | 10 - 11 | 989 | 157 | |
| GenBank mRNA | AK009734.1 | - | 9 - 10 | 868 | 157 | |
| AK016787.1 | - | 10 - 11 | 989 | 157 | ||
| AK034931.1 | - | 655 | 255 | |||
| BC010729.1 | - | 10 - 11 | 982 | 3708143 | 157 | |
More Information
Updated Mapping Information
Set Membership: |
|

Gpr89
Genomic Map