Assay Details
Gene Symbol: Zfp236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177832.3 | NP_808500.2 | 21 - 22 | 3920 | 78 | |
GenBank mRNA | BC138187.1 | - | 21 - 22 | 3920 | 8861206 | 78 |
BC145180.1 | - | 23 - 24 | 4175 | 78 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
