Assay Details
Gene Symbol: Camk2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_177407.4 | NP_803126.1 | 6 - 7 | 558 | 75 | |
GenBank mRNA | AK083245.1 | - | 6 - 7 | 559 | 75 | |
AK122410.2 | - | 6 - 7 | 554 | 75 | ||
BC031745.1 | - | 6 - 7 | 539 | 4506005 | 75 | |
X14836.1 | - | 6 - 7 | 542 | 75 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
