Assay Details
Gene Symbol: Gnal | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010307.3 | NP_034437.1 | 8 - 9 | 950 | 73 | |
NM_177137.5 | NP_796111.2 | 8 - 9 | 1115 | 73 | ||
GenBank mRNA | AK158831.1 | - | 8 - 9 | 951 | 73 | |
AY179169.1 | - | 8 - 9 | 684 | 73 | ||
BC078439.1 | - | 8 - 9 | 1077 | 5685967 | 73 | |
U38503.1 | - | 1 - 2 | 60 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
