Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR1200011079.mm.1 |
NM_011867.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011079.mm.1 |
NM_011867.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011079.mm.1 |
TC1200001595.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011078.mm.1 |
NM_011867.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011078.mm.1 |
NM_011867.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011078.mm.1 |
TC1200001595.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR1200011080.mm.1 |
NM_011867.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011080.mm.1 |
NM_011867.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR1200011080.mm.1 |
TC1200001595.mm.1 |
MTA TC |