Assay Details
| Gene Symbol: Hdac3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010411.2 | NP_034541.2 | 2 - 3 | 214 | 81 | |
| GenBank mRNA | AF074881.1 | - | 2 - 3 | 161 | 81 | |
| AF074882.1 | - | 2 - 3 | 161 | 81 | ||
| AF125536.1 | - | 2 - 3 | 146 | 81 | ||
| BC139300.1 | - | 2 - 3 | 242 | 8862322 | 81 | |
| BC139301.1 | - | 2 - 3 | 242 | 8862323 | 81 | |
| BQ932476.1 | - | 2 - 3 | 226 | 81 | ||
| CJ166955.1 | - | 2 - 3 | 217 | 81 | ||
| JN651901.1 | - | 2 - 3 | 144 | 81 | ||
| JN651902.1 | - | 2 - 3 | 144 | 81 | ||
| JN651903.1 | - | 2 - 3 | 144 | 81 | ||
More Information
Set Membership: |
|

Hdac3
Genomic Map