Assay Details
| Gene Symbol: Txndc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025334.3 | NP_079610.1 | 1 - 2 | 173 | 76 | |
| GenBank mRNA | AK002862.1 | - | 1 - 2 | 160 | 76 | |
| AK003481.1 | - | 1 - 2 | 154 | 76 | ||
| AY548113.1 | - | 1 - 2 | 85 | 76 | ||
| BC006857.1 | - | 1 - 2 | 173 | 2615934 | 76 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txndc12
Genomic Map