Assay Details
| Gene Symbol: Txndc12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025334.3 | NP_079610.1 | 2 - 3 | 237 | 81 | |
| GenBank mRNA | AK002862.1 | - | 2 - 3 | 224 | 81 | |
| AK003481.1 | - | 2 - 3 | 218 | 81 | ||
| AY548113.1 | - | 2 - 3 | 149 | 81 | ||
| BC006857.1 | - | 2 - 3 | 237 | 2615934 | 81 | |
More Information
Updated Mapping Information
Set Membership: |
|

Txndc12
Genomic Map