Assay Details
Gene Symbol: Nrd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_146150.2 | NP_666262.2 | 16 - 17 | 1958 | 112 | |
GenBank mRNA | AK031548.1 | - | 16 - 17 | 1958 | 112 | |
AK050944.1 | - | 3 - 4 | 261 | 112 | ||
AK166470.1 | - | 14 - 15 | 1957 | 112 | ||
BC023786.1 | - | 15 - 16 | 1336 | 112 | ||
BC026832.1 | - | 16 - 17 | 2008 | 4486176 | 112 | |
BC029977.1 | - | 7 - 8 | 594 | 112 | ||
BC036128.1 | - | 16 - 17 | 2076 | 4971791 | 112 | |
U86112.1 | - | 2 - 3 | 206 | 112 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
